Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615124 MIM: 608434 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD13B - ankyrin repeat domain 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152345.4 | Intron | NP_689558.4 | ||||
XM_005257920.4 | Intron | XP_005257977.1 | ||||
XM_011524319.2 | Intron | XP_011522621.1 | ||||
XM_011524320.2 | Intron | XP_011522622.1 | ||||
XM_011524321.2 | Intron | XP_011522623.1 | ||||
XM_017024174.1 | Intron | XP_016879663.1 | ||||
XM_017024175.1 | Intron | XP_016879664.1 | ||||
XM_017024176.1 | Intron | XP_016879665.1 |
GIT1 - GIT ArfGAP 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |