Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616177 MIM: 147520 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDRGK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDRGK1 - DDRGK domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPA - inosine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267623.1 | 286 | Intron | NP_001254552.1 | |||
NM_001324236.1 | 286 | Intron | NP_001311165.1 | |||
NM_001324237.1 | 286 | UTR 5 | NP_001311166.1 | |||
NM_001324238.1 | 286 | Intron | NP_001311167.1 | |||
NM_001324240.1 | 286 | Missense Mutation | ACA,CCA | T,P 32 | NP_001311169.1 | |
NM_033453.3 | 286 | Missense Mutation | ACA,CCA | T,P 32 | NP_258412.1 | |
NM_181493.3 | 286 | Missense Mutation | ACA,CCA | T,P 15 | NP_852470.1 | |
XM_006723564.3 | 286 | Missense Mutation | ACA,CCA | T,P 32 | XP_006723627.1 | |
XM_006723565.3 | 286 | Intron | XP_006723628.1 | |||
XM_011529234.2 | 286 | Missense Mutation | ACA,CCA | T,P 32 | XP_011527536.1 |
Set Membership: |
HapMap |