Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCN - decorin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001920.4 | Intron | NP_001911.1 | ||||
NM_133503.3 | Intron | NP_598010.1 | ||||
NM_133504.3 | Intron | NP_598011.1 | ||||
NM_133505.3 | Intron | NP_598012.1 | ||||
NM_133506.3 | Intron | NP_598013.1 | ||||
NM_133507.3 | Intron | NP_598014.1 | ||||
XM_005268693.1 | Intron | XP_005268750.1 | ||||
XM_006719270.1 | Intron | XP_006719333.1 | ||||
XM_017018917.1 | Intron | XP_016874406.1 |