Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611221 MIM: 610075 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSDMB - gasdermin B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC3C - leucine rich repeat containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORMDL3 - ORMDL sphingolipid biosynthesis regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320801.1 | 1888 | UTR 3 | NP_001307730.1 | |||
NM_001320802.1 | 1888 | UTR 3 | NP_001307731.1 | |||
NM_001320803.1 | 1888 | UTR 3 | NP_001307732.1 | |||
NM_139280.3 | 1888 | UTR 3 | NP_644809.1 | |||
XM_017025345.1 | 1888 | UTR 3 | XP_016880834.1 | |||
XM_017025346.1 | 1888 | UTR 3 | XP_016880835.1 | |||
XM_017025347.1 | 1888 | UTR 3 | XP_016880836.1 | |||
XM_017025348.1 | 1888 | UTR 3 | XP_016880837.1 |