Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 154235 | ||||||||||||||||||||
Literature Links: |
MAK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAK - male germ cell associated kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242385.1 | 3298 | UTR 3 | NP_001229314.1 | |||
NM_001242957.2 | 3298 | UTR 3 | NP_001229886.1 | |||
NM_005906.5 | 3298 | UTR 3 | NP_005897.1 | |||
XM_011514619.2 | 3298 | UTR 3 | XP_011512921.1 | |||
XM_011514620.2 | 3298 | UTR 3 | XP_011512922.1 | |||
XM_011514621.2 | 3298 | Intron | XP_011512923.1 | |||
XM_011514622.2 | 3298 | UTR 3 | XP_011512924.1 | |||
XM_011514624.2 | 3298 | Intron | XP_011512926.1 | |||
XM_017010863.1 | 3298 | UTR 3 | XP_016866352.1 | |||
XM_017010864.1 | 3298 | UTR 3 | XP_016866353.1 | |||
XM_017010865.1 | 3298 | UTR 3 | XP_016866354.1 | |||
XM_017010866.1 | 3298 | Intron | XP_016866355.1 |
TMEM14B - transmembrane protein 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |