Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf63 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf63 - chromosome 11 open reading frame 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024806.3 | Intron | NP_079082.2 | ||||
NM_199124.2 | Intron | NP_954575.1 | ||||
XM_005271680.4 | Intron | XP_005271737.1 | ||||
XM_006718915.2 | Intron | XP_006718978.1 | ||||
XM_011543009.2 | Intron | XP_011541311.1 | ||||
XM_017018338.1 | Intron | XP_016873827.1 | ||||
XM_017018339.1 | Intron | XP_016873828.1 | ||||
XM_017018340.1 | Intron | XP_016873829.1 | ||||
XM_017018341.1 | Intron | XP_016873830.1 | ||||
XM_017018342.1 | Intron | XP_016873831.1 | ||||
XM_017018343.1 | Intron | XP_016873832.1 | ||||
XM_017018344.1 | Intron | XP_016873833.1 |