Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609212 | ||||||||||||||||||||
Literature Links: |
ASRGL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASRGL1 - asparaginase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083926.1 | Intron | NP_001077395.1 | ||||
NM_025080.3 | Intron | NP_079356.3 | ||||
XM_005274305.3 | Intron | XP_005274362.1 | ||||
XM_005274306.2 | Intron | XP_005274363.1 | ||||
XM_011545265.2 | Intron | XP_011543567.1 | ||||
XM_011545266.2 | Intron | XP_011543568.1 | ||||
XM_017018354.1 | Intron | XP_016873843.1 |