Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610561 MIM: 608547 | ||||||||||||||||||||
Literature Links: |
PRSS53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRSS53 - protease, serine 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039503.2 | 2687 | UTR 3 | NP_001034592.1 | |||
XM_011545816.1 | 2687 | Missense Mutation | CAA,CAT | Q,H 534 | XP_011544118.1 | |
XM_011545817.1 | 2687 | UTR 3 | XP_011544119.1 | |||
XM_011545818.2 | 2687 | UTR 3 | XP_011544120.1 | |||
XM_011545819.1 | 2687 | Missense Mutation | CAA,CAT | Q,H 487 | XP_011544121.1 | |
XM_011545820.1 | 2687 | UTR 3 | XP_011544122.1 |
VKORC1 - vitamin K epoxide reductase complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF646 - zinc finger protein 646 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014699.3 | 2687 | Intron | NP_055514.3 | |||
XM_005255710.3 | 2687 | Intron | XP_005255767.1 | |||
XM_005255711.3 | 2687 | Intron | XP_005255768.1 | |||
XM_005255712.3 | 2687 | Intron | XP_005255769.1 | |||
XM_011545990.2 | 2687 | Intron | XP_011544292.1 |
ZNF668 - zinc finger protein 668 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |