Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180630 | ||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316990.1 | Intron | NP_001303919.1 | ||||
NM_138363.2 | Intron | NP_612372.1 | ||||
XM_005257779.3 | Intron | XP_005257836.1 | ||||
XM_005257780.2 | Intron | XP_005257837.1 | ||||
XM_006722166.3 | Intron | XP_006722229.1 | ||||
XM_011525458.2 | Intron | XP_011523760.1 | ||||
XM_017025287.1 | Intron | XP_016880776.1 | ||||
XM_017025288.1 | Intron | XP_016880777.1 |
DDX5 - DEAD-box helicase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320595.1 | Intron | NP_001307524.1 | ||||
NM_001320596.1 | Intron | NP_001307525.1 | ||||
NM_001320597.1 | Intron | NP_001307526.1 | ||||
NM_004396.4 | Intron | NP_004387.1 |
MIR3064 - microRNA 3064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5047 - microRNA 5047 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |