Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGAGATGGAGTAAGATAAGGA[A/T]TGTGTGTTTAGCTTTAATGTCTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612072 MIM: 611974 MIM: 606521 | ||||||||||||||||||||
Literature Links: |
LOC100287042 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100287042 - uncharacterized LOC100287042 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIF4GD - MIF4G domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS7 - mitochondrial ribosomal protein S7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A19 - solute carrier family 25 member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126121.1 | 1840 | UTR 3 | NP_001119593.1 | |||
NM_001126122.1 | 1840 | UTR 3 | NP_001119594.1 | |||
NM_021734.4 | 1840 | UTR 3 | NP_068380.3 | |||
XM_005257559.3 | 1840 | UTR 3 | XP_005257616.1 | |||
XM_005257560.2 | 1840 | UTR 3 | XP_005257617.1 | |||
XM_005257561.3 | 1840 | UTR 3 | XP_005257618.1 | |||
XM_005257562.2 | 1840 | UTR 3 | XP_005257619.1 | |||
XM_006722007.2 | 1840 | UTR 3 | XP_006722070.1 | |||
XM_011525098.1 | 1840 | Intron | XP_011523400.1 | |||
XM_017024926.1 | 1840 | UTR 3 | XP_016880415.1 | |||
XM_017024927.1 | 1840 | UTR 3 | XP_016880416.1 | |||
XM_017024928.1 | 1840 | UTR 3 | XP_016880417.1 |