Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604761 | ||||||||||||||||||||
Literature Links: |
ZFAND5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFAND5 - zinc finger AN1-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102420.2 | 2384 | UTR 3 | NP_001095890.1 | |||
NM_001102421.2 | 2384 | UTR 3 | NP_001095891.1 | |||
NM_001278243.1 | 2384 | UTR 3 | NP_001265172.1 | |||
NM_001278244.1 | 2384 | UTR 3 | NP_001265173.1 | |||
NM_001278245.1 | 2384 | UTR 3 | NP_001265174.1 | |||
NM_006007.3 | 2384 | UTR 3 | NP_005998.1 |