Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601828 | ||||||||||||||||||||
Literature Links: |
NR4A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR4A2 - nuclear receptor subfamily 4 group A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006186.3 | 4637 | UTR 3 | NP_006177.1 | |||
XM_005246621.3 | 4637 | UTR 3 | XP_005246678.1 | |||
XM_005246622.3 | 4637 | UTR 3 | XP_005246679.1 | |||
XM_006712553.3 | 4637 | UTR 3 | XP_006712616.1 | |||
XM_011511246.1 | 4637 | UTR 3 | XP_011509548.1 | |||
XM_017004219.1 | 4637 | UTR 3 | XP_016859708.1 | |||
XM_017004220.1 | 4637 | UTR 3 | XP_016859709.1 |