Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCGCTTGGGCAGCGGCTGTTTG[G/C]GCTTATGCCCCAGCCTGCCCTCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXJ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXJ1 - forkhead box J1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001454.3 | 1176 | Missense Mutation | CCC,CGC | P,R 274 | NP_001445.2 |
RNF157 - ring finger protein 157 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF157-AS1 - RNF157 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |