Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610036 MIM: 610339 | ||||||||||||||||||||
Literature Links: |
CLDN19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLDN19 - claudin 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P3H1 - prolyl 3-hydroxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146289.1 | 2449 | UTR 3 | NP_001139761.1 | |||
NM_001243246.1 | 2449 | UTR 3 | NP_001230175.1 | |||
NM_022356.3 | 2449 | UTR 3 | NP_071751.3 | |||
XM_005271110.2 | 2449 | UTR 3 | XP_005271167.1 | |||
XM_011541948.1 | 2449 | UTR 3 | XP_011540250.1 | |||
XM_017002051.1 | 2449 | UTR 3 | XP_016857540.1 | |||
XM_017002052.1 | 2449 | UTR 3 | XP_016857541.1 |