Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf1 - chromosome 11 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022761.2 | 428 | Missense Mutation | CAC,CAG | H,Q 85 | NP_073598.1 | |
XM_011542957.2 | 428 | Missense Mutation | CAC,CAG | H,Q 101 | XP_011541259.1 | |
XM_011542958.2 | 428 | Missense Mutation | CAC,CAG | H,Q 101 | XP_011541260.1 | |
XM_017018129.1 | 428 | Missense Mutation | CAC,CAG | H,Q 85 | XP_016873618.1 |
FDXACB1 - ferredoxin-fold anticodon binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |