Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147470 | ||||||||||||||||||||
Literature Links: |
IGF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGF2 - insulin like growth factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000612.5 | 4326 | UTR 3 | NP_000603.1 | |||
NM_001007139.5 | 4326 | UTR 3 | NP_001007140.2 | |||
NM_001127598.2 | 4326 | UTR 3 | NP_001121070.1 | |||
NM_001291861.2 | 4326 | UTR 3 | NP_001278790.1 | |||
NM_001291862.2 | 4326 | UTR 3 | NP_001278791.1 |
INS-IGF2 - INS-IGF2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042376.2 | 4326 | Intron | NP_001035835.1 |
MIR483 - microRNA 483 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |