Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602375 MIM: 606794 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MRPL12 - mitochondrial ribosomal protein L12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A10 - solute carrier family 25 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270888.1 | 499 | Silent Mutation | ACC,ACT | T,T 54 | NP_001257817.1 | |
NM_001270953.1 | 499 | Silent Mutation | ACC,ACT | T,T 54 | NP_001257882.1 | |
NM_012140.4 | 499 | Silent Mutation | ACC,ACT | T,T 54 | NP_036272.2 | |
XM_017024220.1 | 499 | Silent Mutation | ACC,ACT | T,T 11 | XP_016879709.1 |
Set Membership: |
HapMap |