Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF529 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF529 - zinc finger protein 529 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145649.1 | 4128 | UTR 3 | NP_001139121.1 | |||
NM_001145650.1 | 4128 | UTR 3 | NP_001139122.1 | |||
NM_001321351.1 | 4128 | UTR 3 | NP_001308280.1 | |||
NM_020951.4 | 4128 | UTR 3 | NP_066002.3 | |||
XM_006723302.3 | 4128 | UTR 3 | XP_006723365.1 | |||
XM_006723304.2 | 4128 | UTR 3 | XP_006723367.1 | |||
XM_011527164.2 | 4128 | UTR 3 | XP_011525466.1 | |||
XM_011527165.2 | 4128 | UTR 3 | XP_011525467.1 | |||
XM_011527166.2 | 4128 | UTR 3 | XP_011525468.1 | |||
XM_011527167.2 | 4128 | UTR 3 | XP_011525469.1 | |||
XM_011527169.2 | 4128 | Intron | XP_011525471.1 | |||
XM_011527170.1 | 4128 | Intron | XP_011525472.1 | |||
XM_017027039.1 | 4128 | UTR 3 | XP_016882528.1 | |||
XM_017027040.1 | 4128 | UTR 3 | XP_016882529.1 | |||
XM_017027041.1 | 4128 | UTR 3 | XP_016882530.1 | |||
XM_017027042.1 | 4128 | UTR 3 | XP_016882531.1 | |||
XM_017027043.1 | 4128 | UTR 3 | XP_016882532.1 | |||
XM_017027044.1 | 4128 | UTR 3 | XP_016882533.1 | |||
XM_017027045.1 | 4128 | UTR 3 | XP_016882534.1 |