Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615417 MIM: 608356 MIM: 609146 MIM: 604481 | ||||||||||||||||||||
Literature Links: |
BET1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BET1L - Bet1 golgi vesicular membrane trafficking protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098787.1 | 650 | UTR 3 | NP_001092257.1 | |||
NM_016526.4 | 650 | UTR 3 | NP_057610.2 |
MIR6743 - microRNA 6743 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF3 - outer dense fiber of sperm tails 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIC8A - RIC8 guanine nucleotide exchange factor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT3 - sirtuin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |