Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611204 MIM: 601404 | ||||||||||||||||||||
Literature Links: |
CCDC88C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC88C - coiled-coil domain containing 88C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080414.3 | 4562 | UTR 3 | NP_001073883.2 | |||
XM_005267691.4 | 4562 | Intron | XP_005267748.1 | |||
XM_011536796.2 | 4562 | UTR 3 | XP_011535098.1 | |||
XM_017021335.1 | 4562 | Intron | XP_016876824.1 | |||
XM_017021336.1 | 4562 | UTR 3 | XP_016876825.1 | |||
XM_017021337.1 | 4562 | Intron | XP_016876826.1 |
GPR68 - G protein-coupled receptor 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |