Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 113530 MIM: 612832 | ||||||||||||||||||||
Literature Links: |
BCAT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAT2 - branched chain amino acid transaminase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSD17B14 - hydroxysteroid 17-beta dehydrogenase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016246.2 | 1092 | UTR 3 | NP_057330.2 | |||
XM_005258969.3 | 1092 | UTR 3 | XP_005259026.1 | |||
XM_005258970.3 | 1092 | UTR 3 | XP_005259027.1 |