Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607409 | ||||||||||||||||||||
Literature Links: |
NRN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRN1 - neuritin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278710.1 | 1468 | Silent Mutation | TTC,TTT | F,F 142 | NP_001265639.1 | |
NM_001278711.1 | 1468 | Silent Mutation | TTC,TTT | F,F 168 | NP_001265640.1 | |
NM_016588.2 | 1468 | Silent Mutation | TTC,TTT | F,F 142 | NP_057672.1 | |
XM_006715106.3 | 1468 | Silent Mutation | TTC,TTT | F,F 203 | XP_006715169.1 | |
XM_006715107.3 | 1468 | Silent Mutation | TTC,TTT | F,F 133 | XP_006715170.1 |