Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCAGCACCCTGCGGGCCCGGGG[C/T]CTGGCAGAGTGGTATGGGCACCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FNDC8 - fibronectin type III domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017559.2 | 1166 | UTR 3 | NP_060029.1 |
NLE1 - notchless homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014445.2 | 1166 | Intron | NP_001014445.1 | |||
NM_018096.4 | 1166 | Intron | NP_060566.2 | |||
XM_017024777.1 | 1166 | UTR 3 | XP_016880266.1 |
RAD51L3-RFFL - RAD51L3-RFFL readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |