Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGTTCTGGGGCGCTTGGCTTCA[C/T]CGCGCAGGCGCGGTCGCCGCTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WRB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WRB - tryptophan rich basic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146218.1 | 61 | Intron | NP_001139690.1 | |||
NM_004627.4 | 61 | UTR 5 | NP_004618.2 |
WRB-SH3BGR - WRB-SH3BGR readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317744.1 | 61 | UTR 5 | NP_001304673.1 |