Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTTATCCTTTGTTCATCGTTTC[A/G]TGGAGGGCCGTGGGGCCACCACCTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615900 | |||||||||||||||||||||||
Literature Links: |
AGBL5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AGBL5 - ATP/GTP binding protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035507.2 | 608 | Missense Mutation | ATG,GTG | M,V 144 | NP_001030584.1 | |
NM_021831.5 | 608 | Missense Mutation | ATG,GTG | M,V 144 | NP_068603.4 | |
XM_005264477.2 | 608 | Missense Mutation | ATG,GTG | M,V 144 | XP_005264534.1 | |
XM_011533011.2 | 608 | Missense Mutation | ATG,GTG | M,V 144 | XP_011531313.1 | |
XM_011533012.2 | 608 | Missense Mutation | ATG,GTG | M,V 144 | XP_011531314.1 | |
XM_011533013.2 | 608 | Missense Mutation | ATG,GTG | M,V 144 | XP_011531315.1 |
AGBL5-AS1 - AGBL5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRA-AGC8-1 - transfer RNA-Ala (AGC) 8-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |