Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609095 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO21 - F-box protein 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015002.2 | 3524 | UTR 3 | NP_055817.1 | |||
NM_033624.2 | 3524 | UTR 3 | NP_296373.1 | |||
XM_017019037.1 | 3524 | UTR 3 | XP_016874526.1 | |||
XM_017019038.1 | 3524 | UTR 3 | XP_016874527.1 | |||
XM_017019039.1 | 3524 | UTR 3 | XP_016874528.1 |
TESC-AS1 - TESC antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |