Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603581 | ||||||||||||||||||||
Literature Links: |
PCDH9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDH9 - protocadherin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318372.1 | 6143 | UTR 3 | NP_001305301.1 | |||
NM_001318373.1 | 6143 | UTR 3 | NP_001305302.1 | |||
NM_001318374.1 | 6143 | Intron | NP_001305303.1 | |||
NM_020403.4 | 6143 | UTR 3 | NP_065136.1 | |||
NM_203487.2 | 6143 | UTR 3 | NP_982354.1 | |||
XM_005266408.3 | 6143 | Intron | XP_005266465.1 | |||
XM_011535099.2 | 6143 | Intron | XP_011533401.1 | |||
XM_017020619.1 | 6143 | UTR 3 | XP_016876108.1 | |||
XM_017020620.1 | 6143 | Intron | XP_016876109.1 | |||
XM_017020621.1 | 6143 | Intron | XP_016876110.1 |