Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA7 - carbonic anhydrase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014435.1 | 77 | Intron | NP_001014435.1 | |||
NM_005182.2 | 77 | UTR 5 | NP_005173.1 | |||
XM_005256136.3 | 77 | Intron | XP_005256193.1 | |||
XM_011523311.1 | 77 | UTR 5 | XP_011521613.1 | |||
XM_011523312.1 | 77 | UTR 5 | XP_011521614.1 |