Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603494 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
URI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
URI1 - URI1, prefoldin like chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252641.1 | 16 | UTR 5 | NP_001239570.1 | |||
NM_003796.3 | 16 | Intron | NP_003787.2 | |||
XM_005259362.2 | 16 | UTR 5 | XP_005259419.1 | |||
XM_005259363.4 | 16 | Intron | XP_005259420.1 | |||
XM_011527435.1 | 16 | Intron | XP_011525737.1 |
Set Membership: |
HapMap |