Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602776 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REV3L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
REV3L - REV3 like, DNA directed polymerase zeta catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286431.1 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2958 | NP_001273360.1 | |
NM_001286432.1 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2958 | NP_001273361.1 | |
NM_002912.4 | 9738 | Silent Mutation | TAC,TAT | Y,Y 3036 | NP_002903.3 | |
XM_011536028.1 | 9738 | Silent Mutation | TAC,TAT | Y,Y 3063 | XP_011534330.1 | |
XM_011536029.2 | 9738 | Silent Mutation | TAC,TAT | Y,Y 3062 | XP_011534331.1 | |
XM_011536030.2 | 9738 | Silent Mutation | TAC,TAT | Y,Y 3037 | XP_011534332.1 | |
XM_011536031.2 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2985 | XP_011534333.1 | |
XM_011536032.2 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2985 | XP_011534334.1 | |
XM_011536036.2 | 9738 | Intron | XP_011534338.1 | |||
XM_017011152.1 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2984 | XP_016866641.1 | |
XM_017011153.1 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2984 | XP_016866642.1 | |
XM_017011154.1 | 9738 | Silent Mutation | TAC,TAT | Y,Y 2984 | XP_016866643.1 | |
XM_017011155.1 | 9738 | Intron | XP_016866644.1 |
Set Membership: |
HapMap |