Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605377 MIM: 600963 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHMG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BHMG1 - basic helix-loop-helix and HMG-box containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DMPK - dystrophia myotonica protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081560.2 | 3075 | UTR 3 | NP_001075029.1 | |||
NM_001081562.2 | 3075 | UTR 3 | NP_001075031.1 | |||
NM_001081563.2 | 3075 | UTR 3 | NP_001075032.1 | |||
NM_001288764.1 | 3075 | UTR 3 | NP_001275693.1 | |||
NM_001288765.1 | 3075 | UTR 3 | NP_001275694.1 | |||
NM_001288766.1 | 3075 | UTR 3 | NP_001275695.1 | |||
NM_004409.4 | 3075 | UTR 3 | NP_004400.4 |
SIX5 - SIX homeobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175875.4 | 3075 | Intron | NP_787071.2 |
Set Membership: |
HapMap |