Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604523 MIM: 610230 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CELSR1 - cadherin EGF LAG seven-pass G-type receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014246.1 | 11058 | UTR 3 | NP_055061.1 | |||
XM_006724383.3 | 11058 | Intron | XP_006724446.1 | |||
XM_011530553.1 | 11058 | Intron | XP_011528855.1 | |||
XM_011530554.2 | 11058 | Intron | XP_011528856.1 | |||
XM_011530555.2 | 11058 | Intron | XP_011528857.1 |
TRMU - tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |