Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGAPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RALGAPA2 - Ral GTPase activating protein catalytic alpha subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020343.3 | 8408 | UTR 3 | NP_065076.2 | |||
XM_005260768.2 | 8408 | Intron | XP_005260825.1 | |||
XM_006723598.1 | 8408 | UTR 3 | XP_006723661.1 | |||
XM_006723599.1 | 8408 | UTR 3 | XP_006723662.1 | |||
XM_011529309.1 | 8408 | Intron | XP_011527611.1 | |||
XM_011529310.2 | 8408 | UTR 3 | XP_011527612.1 | |||
XM_011529311.2 | 8408 | UTR 3 | XP_011527613.2 | |||
XM_017027972.1 | 8408 | Intron | XP_016883461.1 | |||
XM_017027973.1 | 8408 | UTR 3 | XP_016883462.1 | |||
XM_017027974.1 | 8408 | UTR 3 | XP_016883463.1 | |||
XM_017027975.1 | 8408 | UTR 3 | XP_016883464.1 | |||
XM_017027976.1 | 8408 | UTR 3 | XP_016883465.1 | |||
XM_017027977.1 | 8408 | Intron | XP_016883466.1 |
Set Membership: |
HapMap |