Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGKB - diacylglycerol kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004080.2 | 4360 | UTR 3 | NP_004071.1 | |||
NM_145695.2 | 4360 | Intron | NP_663733.1 | |||
XM_005249628.2 | 4360 | UTR 3 | XP_005249685.1 | |||
XM_005249629.2 | 4360 | UTR 3 | XP_005249686.1 | |||
XM_005249630.2 | 4360 | UTR 3 | XP_005249687.1 | |||
XM_005249631.3 | 4360 | UTR 3 | XP_005249688.1 | |||
XM_011515153.2 | 4360 | UTR 3 | XP_011513455.1 | |||
XM_011515154.2 | 4360 | UTR 3 | XP_011513456.1 | |||
XM_011515156.2 | 4360 | UTR 3 | XP_011513458.1 | |||
XM_011515157.2 | 4360 | UTR 3 | XP_011513459.1 | |||
XM_011515158.2 | 4360 | UTR 3 | XP_011513460.1 | |||
XM_017011783.1 | 4360 | UTR 3 | XP_016867272.1 | |||
XM_017011784.1 | 4360 | UTR 3 | XP_016867273.1 | |||
XM_017011785.1 | 4360 | UTR 3 | XP_016867274.1 | |||
XM_017011786.1 | 4360 | UTR 3 | XP_016867275.1 | |||
XM_017011787.1 | 4360 | UTR 3 | XP_016867276.1 | |||
XM_017011788.1 | 4360 | UTR 3 | XP_016867277.1 | |||
XM_017011789.1 | 4360 | UTR 3 | XP_016867278.1 | |||
XM_017011790.1 | 4360 | Intron | XP_016867279.1 | |||
XM_017011791.1 | 4360 | Intron | XP_016867280.1 | |||
XM_017011792.1 | 4360 | Intron | XP_016867281.1 |
Set Membership: |
HapMap |