Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610076 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDK20 - cyclin dependent kinase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039803.2 | 1504 | UTR 3 | NP_001034892.1 | |||
NM_001170639.1 | 1504 | UTR 3 | NP_001164110.1 | |||
NM_001170640.1 | 1504 | UTR 3 | NP_001164111.1 | |||
NM_012119.4 | 1504 | UTR 3 | NP_036251.2 | |||
NM_178432.3 | 1504 | UTR 3 | NP_848519.1 | |||
XM_017014561.1 | 1504 | UTR 3 | XP_016870050.1 | |||
XM_017014562.1 | 1504 | UTR 3 | XP_016870051.1 | |||
XM_017014563.1 | 1504 | UTR 3 | XP_016870052.1 | |||
XM_017014564.1 | 1504 | UTR 3 | XP_016870053.1 |
Set Membership: |
HapMap |