Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609389 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5F - inositol polyphosphate-5-phosphatase F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243194.1 | 2076 | Intron | NP_001230123.1 | |||
NM_001243195.1 | 2076 | Intron | NP_001230124.1 | |||
NM_014937.3 | 2076 | Intron | NP_055752.1 | |||
XM_006717720.3 | 2076 | UTR 5 | XP_006717783.1 | |||
XM_011539524.2 | 2076 | Intron | XP_011537826.1 | |||
XM_011539525.2 | 2076 | UTR 5 | XP_011537827.1 | |||
XM_011539527.2 | 2076 | Intron | XP_011537829.2 | |||
XM_011539528.2 | 2076 | Intron | XP_011537830.1 | |||
XM_011539529.1 | 2076 | Intron | XP_011537831.1 | |||
XM_017015935.1 | 2076 | Intron | XP_016871424.1 | |||
XM_017015936.1 | 2076 | Intron | XP_016871425.1 |