Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606723 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAM1A - ADAM metallopeptidase domain 1A (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPKAPK5 - mitogen-activated protein kinase-activated protein kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003668.3 | 8751 | Intron | NP_003659.2 | |||
NM_139078.2 | 8751 | Intron | NP_620777.1 | |||
XM_017020125.1 | 8751 | Intron | XP_016875614.1 | |||
XM_017020126.1 | 8751 | Intron | XP_016875615.1 | |||
XM_017020127.1 | 8751 | UTR 3 | XP_016875616.1 | |||
XM_017020128.1 | 8751 | UTR 3 | XP_016875617.1 | |||
XM_017020129.1 | 8751 | Intron | XP_016875618.1 | |||
XM_017020130.1 | 8751 | Intron | XP_016875619.1 | |||
XM_017020131.1 | 8751 | Intron | XP_016875620.1 | |||
XM_017020132.1 | 8751 | UTR 3 | XP_016875621.1 | |||
XM_017020133.1 | 8751 | UTR 3 | XP_016875622.1 | |||
XM_017020134.1 | 8751 | UTR 3 | XP_016875623.1 | |||
XM_017020135.1 | 8751 | UTR 3 | XP_016875624.1 | |||
XM_017020136.1 | 8751 | UTR 3 | XP_016875625.1 | |||
XM_017020137.1 | 8751 | Intron | XP_016875626.1 | |||
XM_017020138.1 | 8751 | UTR 3 | XP_016875627.1 | |||
XM_017020139.1 | 8751 | Intron | XP_016875628.1 |
Set Membership: |
HapMap |