Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188250 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TK2 - thymidine kinase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172643.1 | 3144 | UTR 3 | NP_001166114.1 | |||
NM_001172644.1 | 3144 | UTR 3 | NP_001166115.1 | |||
NM_001172645.1 | 3144 | UTR 3 | NP_001166116.1 | |||
NM_001271934.1 | 3144 | UTR 3 | NP_001258863.1 | |||
NM_001271935.1 | 3144 | UTR 3 | NP_001258864.1 | |||
NM_001272050.1 | 3144 | UTR 3 | NP_001258979.1 | |||
NM_004614.4 | 3144 | UTR 3 | NP_004605.4 |
Set Membership: |
HapMap |