Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL12RB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL12RB1 - interleukin 12 receptor subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290023.1 | 2568 | UTR 3 | NP_001276952.1 | |||
NM_001290024.1 | 2568 | UTR 3 | NP_001276953.1 | |||
NM_005535.2 | 2568 | UTR 3 | NP_005526.1 | |||
NM_153701.2 | 2568 | Intron | NP_714912.1 | |||
XM_006722741.3 | 2568 | UTR 3 | XP_006722804.2 | |||
XM_011527966.2 | 2568 | UTR 3 | XP_011526268.1 | |||
XM_011527967.2 | 2568 | UTR 3 | XP_011526269.1 | |||
XM_011527968.2 | 2568 | UTR 3 | XP_011526270.1 | |||
XM_011527969.2 | 2568 | UTR 3 | XP_011526271.1 | |||
XM_011527970.2 | 2568 | UTR 3 | XP_011526272.1 | |||
XM_011527971.2 | 2568 | UTR 3 | XP_011526273.1 | |||
XM_011527972.2 | 2568 | UTR 3 | XP_011526274.1 | |||
XM_011527973.2 | 2568 | UTR 3 | XP_011526275.1 | |||
XM_011527974.2 | 2568 | UTR 3 | XP_011526276.1 | |||
XM_011527975.2 | 2568 | UTR 3 | XP_011526277.1 | |||
XM_011527976.2 | 2568 | Intron | XP_011526278.1 | |||
XM_011527977.2 | 2568 | Intron | XP_011526279.1 | |||
XM_017026762.1 | 2568 | UTR 3 | XP_016882251.1 |