Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610924 MIM: 611663 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBCK1 - RANBP2-type and C3HC4-type zinc finger containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D20 - TBC1 domain family member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144628.3 | 1211 | UTR 3 | NP_653229.1 | |||
XM_005260661.1 | 1211 | UTR 3 | XP_005260718.1 | |||
XM_006723540.3 | 1211 | UTR 3 | XP_006723603.1 | |||
XM_017027645.1 | 1211 | UTR 3 | XP_016883134.1 |
Set Membership: |
HapMap |