Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP51 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP51 - ubiquitin specific peptidase 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201286.3 | 634 | Silent Mutation | GAA,GAG | E,E 185 | NP_958443.1 | |
XM_017029299.1 | 634 | Silent Mutation | GAA,GAG | E,E 185 | XP_016884788.1 | |
XM_017029300.1 | 634 | Silent Mutation | GAA,GAG | E,E 185 | XP_016884789.1 | |
XM_017029301.1 | 634 | Silent Mutation | GAA,GAG | E,E 185 | XP_016884790.1 |
Set Membership: |
HapMap |