Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300231 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMGT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MMGT1 - membrane magnesium transporter 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A6 - solute carrier family 9 member A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042537.1 | Intron | NP_001036002.1 | ||||
NM_001177651.1 | Intron | NP_001171122.1 | ||||
NM_006359.2 | Intron | NP_006350.1 | ||||
XM_006724726.3 | Intron | XP_006724789.1 | ||||
XM_011531243.2 | Intron | XP_011529545.1 | ||||
XM_017029223.1 | Intron | XP_016884712.1 | ||||
XM_017029224.1 | Intron | XP_016884713.1 | ||||
XM_017029225.1 | Intron | XP_016884714.1 |
Set Membership: |
HapMap |