Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTGTTTATAAAACTAAGTAAG[C/T]TTCTAAAACAGTGCCTTAAGCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610526 MIM: 616796 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5C1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NT5C1B - 5'-nucleotidase, cytosolic IB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002006.2 | 2198 | UTR 3 | NP_001002006.1 | |||
NM_001199086.1 | 2198 | UTR 3 | NP_001186015.1 | |||
NM_001199087.1 | 2198 | UTR 3 | NP_001186016.1 | |||
NM_001199088.1 | 2198 | UTR 3 | NP_001186017.1 | |||
NM_033253.3 | 2198 | UTR 3 | NP_150278.2 |
NT5C1B-RDH14 - NT5C1B-RDH14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199103.1 | 2198 | Intron | NP_001186032.1 | |||
NM_001199104.1 | 2198 | Intron | NP_001186033.1 |
RDH14 - retinol dehydrogenase 14 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |