Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606879 | ||||||||||||||||||||
Literature Links: |
PHGDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHGDH - phosphoglycerate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006623.3 | 223 | UTR 5 | NP_006614.2 | |||
XM_011541226.1 | 223 | UTR 5 | XP_011539528.1 | |||
XM_011541227.2 | 223 | Intron | XP_011539529.1 | |||
XM_011541228.1 | 223 | Intron | XP_011539530.1 | |||
XM_011541231.2 | 223 | UTR 5 | XP_011539533.1 |
Set Membership: |
JSNP |