Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612414 MIM: 164009 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRTOMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRTOMT - leucine rich transmembrane and O-methyltransferase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145307.4 | Intron | NP_001138779.1 | ||||
NM_001145308.4 | Intron | NP_001138780.1 | ||||
NM_001145309.3 | Intron | NP_001138781.1 | ||||
NM_001145310.3 | Intron | NP_001138782.1 | ||||
NM_001205138.3 | Intron | NP_001192067.1 | ||||
NM_001271471.2 | Intron | NP_001258400.1 | ||||
NM_001318803.1 | Intron | NP_001305732.1 | ||||
NM_145309.5 | Intron | NP_660352.1 | ||||
XM_006718473.3 | Intron | XP_006718536.1 | ||||
XM_006718474.3 | Intron | XP_006718537.1 | ||||
XM_011544847.2 | Intron | XP_011543149.1 | ||||
XM_011544848.2 | Intron | XP_011543150.1 | ||||
XM_017017356.1 | Intron | XP_016872845.1 | ||||
XM_017017357.1 | Intron | XP_016872846.1 | ||||
XM_017017358.1 | Intron | XP_016872847.1 | ||||
XM_017017359.1 | Intron | XP_016872848.1 | ||||
XM_017017360.1 | Intron | XP_016872849.1 |
MIR3165 - microRNA 3165 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUMA1 - nuclear mitotic apparatus protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286561.1 | Intron | NP_001273490.1 | ||||
NM_006185.3 | Intron | NP_006176.2 | ||||
XM_006718564.1 | Intron | XP_006718627.1 | ||||
XM_011545054.1 | Intron | XP_011543356.1 | ||||
XM_011545055.1 | Intron | XP_011543357.1 | ||||
XM_011545056.2 | Intron | XP_011543358.1 | ||||
XM_011545057.2 | Intron | XP_011543359.1 | ||||
XM_011545058.1 | Intron | XP_011543360.1 | ||||
XM_011545059.2 | Intron | XP_011543361.1 | ||||
XM_011545060.2 | Intron | XP_011543362.1 | ||||
XM_011545061.2 | Intron | XP_011543363.1 | ||||
XM_011545062.2 | Intron | XP_011543364.1 | ||||
XM_011545063.2 | Intron | XP_011543365.1 | ||||
XM_011545064.1 | Intron | XP_011543366.1 | ||||
XM_011545065.1 | Intron | XP_011543367.1 | ||||
XM_011545066.1 | Intron | XP_011543368.1 | ||||
XM_017017830.1 | Intron | XP_016873319.1 | ||||
XM_017017831.1 | Intron | XP_016873320.1 |
Set Membership: |
HapMap |