Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603427 MIM: 604097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PER3 - period circadian clock 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTS2 - urotensin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006786.3 | Intron | NP_006777.1 | ||||
NM_021995.2 | Intron | NP_068835.1 | ||||
XM_011540537.2 | Intron | XP_011538839.1 | ||||
XM_011540538.1 | Intron | XP_011538840.1 | ||||
XM_017000116.1 | Intron | XP_016855605.1 | ||||
XM_017000117.1 | Intron | XP_016855606.1 | ||||
XM_017000118.1 | Intron | XP_016855607.1 | ||||
XM_017000119.1 | Intron | XP_016855608.1 |