Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603139 MIM: 600822 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK6 - adenylate kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015891.1 | 98 | UTR 5 | NP_001015891.1 | |||
NM_016283.4 | 98 | Intron | NP_057367.1 |
RAD17 - RAD17 checkpoint clamp loader component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278622.1 | 98 | Intron | NP_001265551.1 | |||
NM_002873.1 | 98 | Intron | NP_002864.1 | |||
NM_133338.2 | 98 | UTR 5 | NP_579916.1 | |||
NM_133339.2 | 98 | Intron | NP_579917.1 | |||
NM_133340.2 | 98 | Intron | NP_579918.1 | |||
NM_133341.2 | 98 | Intron | NP_579919.1 | |||
NM_133342.2 | 98 | UTR 5 | NP_579920.1 | |||
NM_133343.1 | 98 | Intron | NP_579921.1 | |||
NM_133344.2 | 98 | UTR 5 | NP_579922.1 | |||
XM_017009679.1 | 98 | UTR 5 | XP_016865168.1 | |||
XM_017009680.1 | 98 | Intron | XP_016865169.1 | |||
XM_017009681.1 | 98 | Intron | XP_016865170.1 |
TAF9 - TATA-box binding protein associated factor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015892.1 | 98 | UTR 5 | NP_001015892.1 | |||
NM_003187.4 | 98 | Intron | NP_003178.1 |
Set Membership: |
HapMap |