Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616929 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTERF2 - mitochondrial transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033050.2 | Intron | NP_001028222.1 | ||||
NM_025198.4 | Intron | NP_079474.2 | ||||
XM_011538767.2 | Intron | XP_011537069.1 | ||||
XM_017019985.1 | Intron | XP_016875474.1 | ||||
XM_017019986.1 | Intron | XP_016875475.1 | ||||
XM_017019987.1 | Intron | XP_016875476.1 |
TMEM263 - transmembrane protein 263 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |