Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605153 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAMP1 - receptor activity modifying protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308353.1 | Intron | NP_001295282.1 | ||||
NM_005855.3 | Intron | NP_005846.1 | ||||
XM_017003152.1 | Intron | XP_016858641.1 | ||||
XM_017003153.1 | Intron | XP_016858642.1 | ||||
XM_017003154.1 | Intron | XP_016858643.1 | ||||
XM_017003155.1 | Intron | XP_016858644.1 | ||||
XM_017003156.1 | Intron | XP_016858645.1 |