Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605363 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GAD1 - glutamate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000817.2 | Intron | NP_000808.2 | ||||
NM_013445.3 | Intron | NP_038473.2 | ||||
XM_005246444.2 | Intron | XP_005246501.1 | ||||
XM_011510922.1 | Intron | XP_011509224.1 | ||||
XM_017003756.1 | Intron | XP_016859245.1 | ||||
XM_017003757.1 | Intron | XP_016859246.1 | ||||
XM_017003758.1 | Intron | XP_016859247.1 |
Set Membership: |
HapMap |